Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006188.1 | NP_001006188.1 | 3 - 4 | 671 | 119 | |
XM_015275281.2 | XP_015130767.1 | 3 - 4 | 745 | 119 | ||
XM_025155454.1 | XP_025011222.1 | 3 - 4 | 650 | 119 | ||
GenBank mRNA | AJ719798.1 | - | 3 - 4 | 671 | 119 | |
BX933911.2 | - | 3 - 4 | 653 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |