Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006232.1 | NP_001006232.1 | 2 - 3 | 200 | 89 | |
GenBank mRNA | AJ719544.1 | - | 2 - 3 | 200 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |