Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006285.1 | NP_001006285.1 | 9 - 10 | 782 | 63 | |
XM_015296246.2 | XP_015151732.1 | 9 - 10 | 884 | 63 | ||
GenBank mRNA | AJ720641.1 | - | 9 - 10 | 782 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |