Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007081.1 | NP_001007082.1 | 5 - 6 | 828 | 68 | |
GenBank mRNA | AY702438.1 | - | 5 - 6 | 828 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |