Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007832.1 | NP_001007833.1 | 2 - 3 | 283 | 95 | |
XM_025154840.1 | XP_025010608.1 | 2 - 3 | 492 | 95 | ||
GenBank mRNA | AJ720358.1 | - | 2 - 3 | 283 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |