Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204742.2 | NP_990073.1 | 7 - 8 | 1248 | 70 | |
GenBank mRNA | AF192787.1 | - | 7 - 8 | 1182 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |