Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250768.1 | NP_001237697.1 | 20 - 21 | 3540 | 87 | |
GenBank mRNA | AM261476.1 | - | 20 - 21 | 3540 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Glycine max Probe spans exons |