Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248786.1 | NP_001235715.1 | 1 | 751 | 62 | |
GenBank mRNA | DQ822890.1 | - | 1 | 751 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |