Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU019592.1 | - | 1 - 2 | 407 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Glycine max Probe spans exons |