Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020300.hg.1 |
TC02001311.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020300.hg.1 |
ENST00000258415.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020300.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010811.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000258415.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000411688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000445971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000466602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
ENST00000494263.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
NM_000784.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177146.hg.1 |
TC0200010811.hg.1 |
Clariom D TC |