Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010098.hg.1 |
TC09000778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010098.hg.1 |
ENST00000371880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010098.hg.1 |
ENST00000393056.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010098.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161945.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161945.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161945.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161945.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161945.hg.1 |
TC0900009089.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000263611.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
ENST00000393056.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009089.hg.1 |
NM_000787.3 |
Refseq |