Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020447.hg.1 |
TC08001552.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020447.hg.1 |
ENST00000297350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020447.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020444.hg.1 |
TC08001552.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020444.hg.1 |
ENST00000297350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020444.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020445.hg.1 |
TC08001552.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020445.hg.1 |
ENST00000297350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020445.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020446.hg.1 |
TC08001552.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020446.hg.1 |
ENST00000297350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020446.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178796.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178795.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178798.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178798.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178798.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178798.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178798.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178797.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800178799.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178799.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178799.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800178799.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800178799.hg.1 |
TC0800011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000297350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000517352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
ENST00000521597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011611.hg.1 |
NM_002546.3 |
Refseq |