Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000286758.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
ENST00000506590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007918.hg.1 |
NM_006419.2 |
Refseq |