Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
ENST00000228945.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
ENST00000541546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
ENST00000541644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016647.hg.1 |
TC12001278.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175733.hg.1 |
TC1200010024.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000228945.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000535676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000536592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000539131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541546.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000541644.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000542276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
ENST00000545895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010024.hg.1 |
NM_001175.6 |
Refseq |