Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023371.hg.1 |
TC01001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023371.hg.1 |
ENST00000340030.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023371.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023371.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
ENST00000340030.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
ENST00000367721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
NM_000639.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
NM_001302746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178237.hg.1 |
TC0100010625.hg.1 |
Clariom D TC |