Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
TC05000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
ENST00000320280.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
ENST00000339292.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
ENST00000506858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
ENST00000508822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
ENST00000509998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005401.hg.1 |
NM_004866.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
TC05000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
ENST00000320280.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
ENST00000339292.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
ENST00000506858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
ENST00000508822.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
ENST00000509998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005402.hg.1 |
NM_004866.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000320280.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000506858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000614488.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000618166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000621999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NM_001290229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NM_004866.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NR_110885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000320280.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000506858.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000614488.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000618166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
ENST00000621999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NM_001290229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NM_004866.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
NR_110885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153856.hg.1 |
TC0500007868.hg.1 |
Clariom D TC |