Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007753.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007753.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007753.hg.1 |
ENST00000544348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007753.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007753.hg.1 |
TC11000623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007755.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007755.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007755.hg.1 |
ENST00000544348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007755.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007755.hg.1 |
TC11000623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007754.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007754.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007754.hg.1 |
ENST00000544348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007754.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007754.hg.1 |
TC11000623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007748.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007748.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007748.hg.1 |
TC11000623.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007749.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007749.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007749.hg.1 |
TC11000623.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100157775.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157775.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157775.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157775.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157775.hg.1 |
TC1100008010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157779.hg.1 |
TC1100008010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157778.hg.1 |
TC1100008010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157776.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157776.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157776.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157776.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157776.hg.1 |
TC1100008010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
ENST00000279249.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
ENST00000533419.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
NM_006779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157780.hg.1 |
TC1100008010.hg.1 |
Clariom D TC |