Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017840.hg.1 |
TC0X001327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017840.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017840.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172610.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172610.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172610.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172610.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172610.hg.1 |
TC0X00010678.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000304661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
ENST00000371313.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_001011551.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010678.hg.1 |
NM_152692.4 |
Refseq |