Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003215.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003215.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003215.hg.1 |
TC22000169.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003214.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003214.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003214.hg.1 |
TC22000169.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003216.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003216.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003216.hg.1 |
TC22000169.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148304.hg.1 |
TC2200006947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148304.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148304.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148304.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148304.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148306.hg.1 |
TC2200006947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148306.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148306.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148306.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148306.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148303.hg.1 |
TC2200006947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148303.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148303.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148303.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148303.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148305.hg.1 |
TC2200006947.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200148305.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148305.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200148305.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200148305.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
ENST00000215906.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006947.hg.1 |
NM_020437.4 |
Refseq |