Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020135.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020135.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020135.hg.1 |
TC10001569.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020137.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020137.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020137.hg.1 |
TC10001569.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020140.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020140.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020140.hg.1 |
TC10001569.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000178743.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178743.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178743.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178743.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178743.hg.1 |
TC1000011556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000178745.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178745.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178745.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178745.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178745.hg.1 |
TC1000011556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011556.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178740.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178740.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178740.hg.1 |
ENST00000370626.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178740.hg.1 |
NM_021732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178740.hg.1 |
TC1000011556.hg.1 |
Clariom D TC |