Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026715.hg.1 |
ENST00000299855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026715.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026715.hg.1 |
TC11002235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026718.hg.1 |
ENST00000299855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026718.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026718.hg.1 |
TC11002235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026717.hg.1 |
ENST00000299855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026717.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026717.hg.1 |
TC11002235.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026716.hg.1 |
ENST00000299855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026716.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026716.hg.1 |
TC11002235.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193281.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193282.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193283.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193285.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193285.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193285.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193285.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193285.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193286.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193280.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193287.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193288.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |