Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343124 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343116 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343143 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184521 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343138 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343130 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343102 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343120 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184528 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184500 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184484 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184489 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343113 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343134 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184486 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343129 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184544 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343119 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184533 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343150 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343145 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184502 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343104 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343127 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343158 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184492 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343107 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184497 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343132 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343151 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343122 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184541 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184494 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184491 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343139 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343156 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343137 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343141 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343140 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343135 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184504 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343117 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23343154 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184512 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P03184487 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
47855 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
29712 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P121596 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P121596 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
12719 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P108762 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P108762 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
21020 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
19169 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
13792 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
38695 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
41498 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
50811 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
3650 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
137 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
3582 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
41810 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31298977 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P16728299 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P121596 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_19_P00802024 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P121596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343137 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343135 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343141 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184504 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184535 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343147 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184528 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184497 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184492 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343132 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184544 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343134 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184494 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343113 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184489 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343139 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343119 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343127 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343120 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343129 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184487 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343143 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343122 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343102 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343146 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184538 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343107 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343152 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184500 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184510 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343144 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343147 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184535 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343119 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343141 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343113 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343137 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343102 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343129 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343132 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343152 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184510 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184544 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184500 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343154 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343144 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343107 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184528 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184487 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184489 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184538 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343156 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343134 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343135 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184492 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343158 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184504 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343127 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343143 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343122 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184494 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343120 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343139 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P03184497 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23343146 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P121596 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P108762 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184489 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184533 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184530 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184531 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184525 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184503 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184509 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184496 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184514 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184499 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184543 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184536 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184511 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184527 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184545 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184519 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184521 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184500 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184540 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184516 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184501 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184507 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184523 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184505 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184483 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184487 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P03184494 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P108762 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P108762 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P121596 |
- |
- |
Custom Human Agilent Array 11k, Chromosome 6, Expression |
1464 |
- |
- |
Custom Human Agilent Array 11k, Chromosome 6, Expression |
7185 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
AJ314847.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
AK022638.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
AK095497.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
AK291282.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
BC002612.2 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
NM_001039885.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
178958 |
NM_024301.3 |
refseq_mrna_human_NM |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
63043 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P108762 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184441 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184468 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184459 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184535 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184565 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184525 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184516 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184530 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184487 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184461 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184449 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184533 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184558 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184517 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184509 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184505 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184543 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184467 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184547 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184450 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184496 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184561 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184538 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184503 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184491 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184483 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184500 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184437 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184457 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184563 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184448 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184555 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184501 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184549 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15582238 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184446 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184523 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184456 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184540 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184444 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184519 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184551 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184545 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15582236 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184536 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184440 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184514 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184511 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184507 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184435 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184521 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184471 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15582244 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184494 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184463 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P03184499 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P108762 |
- |
- |