Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034844.hg.1 |
TC01002286.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034844.hg.1 |
ENST00000375499.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034844.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196629.hg.1 |
TC0100013073.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000375499.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000463045.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000466613.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000475506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000485515.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
ENST00000491274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013073.hg.1 |
NM_003000.2 |
Refseq |