Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
TC03000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
ENST00000243253.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
ENST00000464451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
ENST00000483956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012069.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012070.hg.1 |
TC03000670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012070.hg.1 |
ENST00000243253.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012070.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012070.hg.1 |
ENST00000464451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012070.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000243253.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000464451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000243253.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
ENST00000464451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161957.hg.1 |
TC0300008713.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000243253.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000464451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000483956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000243253.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000424880.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000464451.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
ENST00000483956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
NM_013336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300161956.hg.1 |
TC0300008713.hg.1 |
Clariom D TC |