Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294340.1 | NP_001281269.1 | 14 - 15 | 2383 | 89 | |
NM_144604.3 | NP_653205.3 | 13 - 14 | 2311 | 89 | ||
XM_011522863.1 | XP_011521165.1 | 12 - 13 | 1996 | 89 | ||
XM_011522865.1 | XP_011521167.1 | 6 - 7 | 753 | 89 | ||
GenBank mRNA | AK091849.1 | - | 8 - 9 | 1138 | 89 | |
AK225807.1 | - | 13 - 14 | 2281 | 89 | ||
AK300254.1 | - | 14 - 15 | 2361 | 89 | ||
BC001584.1 | - | 10 - 11 | 1407 | 89 | ||
BC050463.1 | - | 13 - 14 | 2258 | 6042036 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |