Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016144.hg.1 |
ENST00000322347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016144.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016144.hg.1 |
TC15001652.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016145.hg.1 |
ENST00000322347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016145.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15016145.hg.1 |
TC15001652.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174969.hg.1 |
TC1500010050.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174967.hg.1 |
TC1500010050.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000322347.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000562233.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000564811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000567270.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
ENST00000568783.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010050.hg.1 |
NM_004255.3 |
Refseq |