Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_024886_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7936024 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7936026 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7936027 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7936025 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7936023 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
220152_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11736824_at |
- |
- |
Affymetrix Human Genome U219 Array |
11736824_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021134.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021134.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021134.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021133.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021133.hg.1 |
ENST00000473970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021133.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021133.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021136.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021136.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021136.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021135.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021135.hg.1 |
ENST00000473970.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021135.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021135.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021138.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021138.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021138.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021137.hg.1 |
ENST00000239125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021137.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10021137.hg.1 |
TC10001622.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000179984.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179984.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179984.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179984.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179984.hg.1 |
TC1000011681.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000179983.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179983.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179983.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179983.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179983.hg.1 |
TC1000011681.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000473970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000492465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000494270.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000594818.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000596366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000597488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
ENST00000598368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038937.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008731.hg.1 |
NR_038938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
ENST00000625129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011681.hg.1 |
NM_024886.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
213964_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
220152_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
213964_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
220152_at |
- |
- |
Human Genome U133A 2.0 Array |
220152_at |
- |
- |
Human Genome U133A 2.0 Array |
220152_PM_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_024886_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw536267 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
220152_at3 |
- |
- |
Human Genome U95 Set |
51733_at |
- |
- |
Human Genome U95 Set |
51733_PM_at |
- |
- |
Human Genome U95 Set |
73224_PM_r_at |
- |
- |
Human Genome U95 Set |
73224_r_at |
- |
- |
Human Genome U133 Set |
220152_at |
- |
- |
Human Genome U133 Set |
220152_PM_at |
- |
- |