Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132085 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132097 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132114 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779280 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779275 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779244 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132110 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779293 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132142 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132153 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779291 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132090 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132108 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132120 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132087 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132074 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132083 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132132 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779283 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132095 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132155 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132080 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779249 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132151 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132126 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132067 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779273 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779265 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132093 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132073 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132082 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132122 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132129 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779248 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132070 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132099 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779256 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779262 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132077 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132139 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132128 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779229 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132071 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132134 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779281 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132075 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132084 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779269 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132072 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779252 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132148 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132069 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132086 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132103 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779236 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779274 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779221 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779232 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779282 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132149 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132125 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132117 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132115 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132137 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132098 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132138 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132089 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132140 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P31779261 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P11132118 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
AK093925.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
AK312756.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
BC004279.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
BC035665.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
CR623806.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
NM_002155.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
167440 |
X51758.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
41554 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
266837 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
836789 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P166159 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P166159 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
38985 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P107113 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P107113 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
22220 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P22381000 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P34682980 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P41304679 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P03506030 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P03506035 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P166159 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P166159 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779256 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132067 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132128 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132070 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132129 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132137 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132142 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132108 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779283 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132118 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132155 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779252 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779269 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132114 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132121 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132122 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132135 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132110 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779275 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132139 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779282 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132126 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779262 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779286 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779291 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779249 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132151 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779293 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132115 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132149 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132153 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779253 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132122 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132114 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132149 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779248 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132142 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779261 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132128 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132070 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779293 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132151 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779281 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779291 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132126 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132067 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779253 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132153 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132140 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779249 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779273 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132115 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132121 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132129 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132139 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779262 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779256 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779283 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779282 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779275 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779286 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132117 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132137 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132069 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779252 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132135 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132148 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132118 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132155 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132110 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P11132108 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P31779269 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P166159 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P107113 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132115 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132130 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132108 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132126 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132143 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132139 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132122 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132111 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132120 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132155 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132128 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132146 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132157 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132135 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132150 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132147 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132116 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132117 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132136 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132153 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132149 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132121 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P11132137 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P11132122 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P11132120 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P11132119 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P11132121 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17186663 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P11132123 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17186664 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P17186665 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P107113 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P107113 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P166159 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
47513 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
88425 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P107113 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03506035 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P03506032 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132097 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132099 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132155 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132149 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132101 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132111 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132083 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132122 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132136 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132093 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132146 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132077 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132153 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132121 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132115 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132087 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132140 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132080 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132126 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132070 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132116 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132073 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132095 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132143 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132075 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132078 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132151 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132072 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132084 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132135 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132105 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132120 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132103 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132159 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132124 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132130 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132091 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132108 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P11132128 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P03506035 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P107113 |
- |
- |