Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302769.1 | NP_001289698.1 | 11 - 12 | 1829 | 126 | |
NM_057177.6 | NP_476518.4 | 11 - 12 | 1829 | 126 | ||
NM_205863.3 | NP_995585.2 | 11 - 12 | 1829 | 126 | ||
XM_011510552.2 | XP_011508854.1 | 14 - 15 | 2172 | 126 | ||
XM_011510553.2 | XP_011508855.1 | 13 - 14 | 6895 | 126 | ||
XM_017003283.1 | XP_016858772.1 | 10 - 11 | 2119 | 126 | ||
XM_017003284.1 | XP_016858773.1 | 11 - 12 | 5165 | 126 | ||
XM_017003286.1 | XP_016858775.1 | 10 - 11 | 5061 | 126 | ||
XM_017003287.1 | XP_016858776.1 | 13 - 14 | 6898 | 126 | ||
XM_017003288.1 | XP_016858777.1 | 13 - 14 | 6900 | 126 | ||
XM_017003289.1 | XP_016858778.1 | 8 - 9 | 1276 | 126 | ||
XM_017003290.1 | XP_016858779.1 | 12 - 13 | 1776 | 126 | ||
XM_017003291.1 | XP_016858780.1 | 11 - 12 | 1648 | 126 | ||
XM_017003292.1 | XP_016858781.1 | 11 - 12 | 1710 | 126 | ||
XM_017003293.1 | XP_016858782.1 | 8 - 9 | 1214 | 126 | ||
XM_017003294.1 | XP_016858783.1 | 13 - 14 | 6892 | 126 | ||
GenBank mRNA | AB073472.1 | - | 13 - 14 | 1622 | 126 | |
AB092439.1 | - | 13 - 14 | 1622 | 126 | ||
AF428250.1 | - | 11 - 12 | 1622 | 126 | ||
AF466152.1 | - | 11 - 12 | 1622 | 126 | ||
AK056157.1 | - | 13 - 14 | 1710 | 125 | ||
AK057965.1 | - | 6 - 7 | 987 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |