Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014148.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014148.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014148.hg.1 |
TC20001034.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000168535.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168535.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168535.hg.1 |
TC2000009817.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168535.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168535.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
ENST00000217188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009817.hg.1 |
NM_080823.3 |
Refseq |