Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027133.hg.1 |
TC12003284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027133.hg.1 |
ENST00000267205.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027133.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027132.hg.1 |
TC12003284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027132.hg.1 |
ENST00000267205.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12027132.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204633.hg.1 |
TC1200012859.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204636.hg.1 |
TC1200012859.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204635.hg.1 |
TC1200012859.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000267205.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000535560.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
ENST00000537265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012859.hg.1 |
NM_019034.2 |
Refseq |