Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058219.2 | NP_478126.1 | 1 | 184 | 100 | |
GenBank mRNA | BC052252.1 | - | 1 | 184 | 6256140 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10095142 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10095150 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10095145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30636415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30636432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16909955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10095146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30636422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30636397 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 210563 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P39639 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P39963 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P39639 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P121794 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P100413 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 27220 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 27334 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55150 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P33961914 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P39639 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P39639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30636432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095145 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10095145 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30636432 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P121794 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P39639 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P100413 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P39963 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10095146 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10095147 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10095145 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10095148 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10095150 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909980 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909979 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10095145 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909984 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909977 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909983 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909978 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909981 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10095146 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10095147 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16909982 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P39639 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P100413 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33961914 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 588789 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 588789 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095146 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095150 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095142 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16909979 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16909983 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16909981 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095148 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095147 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10095145 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_058219_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8002346 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8002345 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8002344 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 231916_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553947_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1558044_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11735184_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735184_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017026.hg.1 | ENST00000435634.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017026.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017026.hg.1 | TC16001233.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600183087.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183087.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183087.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183087.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183087.hg.1 | TC1600010751.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600183086.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183086.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183086.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183086.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183086.hg.1 | TC1600010751.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | ENST00000435634.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010751.hg.1 | NM_058219.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 231916_at | - | - |
Human Genome U133 Plus 2.0 Array | 227696_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553947_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553947_at | - | - |
Human Genome U133 Plus 2.0 Array | 231916_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 227696_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558044_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1558044_PM_s_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_058219_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553947_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1558044_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231916_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227696_at8 | - | - |
Human Genome U95 Set | 56286_at | - | - |
Human Genome U95 Set | 56286_PM_at | - | - |
Human Genome U95 Set | 63765_PM_r_at | - | - |
Human Genome U95 Set | 63765_r_at | - | - |
Human Genome U95 Set | 63764_i_at | - | - |
Human Genome U95 Set | 63764_PM_i_at | - | - |
Human Genome U133 Set | 231916_PM_at | - | - |
Human Genome U133 Set | 227696_PM_at | - | - |
Human Genome U133 Set | 231916_at | - | - |
Human Genome U133 Set | 227696_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053890 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053890 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 146084 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1670218 | BC052252.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1670218 | AK095542.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_32484988-S | BC052252.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_32484988-S | AK095542.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_32484988-S | NM_058219.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_15247-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003170497 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003170497 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 70477 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_15247 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1670218 | NM_058219.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1670218 | BC052252.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1670218 | AK095542.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1670218 | NM_058219.2 | human_transcriptome |