Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138295.3 | NP_612152.1 | 44 - 45 | 6770 | 75 | |
XM_017011798.1 | XP_016867287.1 | 45 - 46 | 13175 | 75 | ||
GenBank mRNA | AB061683.1 | - | 44 - 45 | 6736 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Homo sapiens Probe spans exons |