Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007607.hg.1 |
ENST00000322172.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007607.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007607.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007607.hg.1 |
TC17000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007609.hg.1 |
ENST00000322172.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007609.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007609.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007609.hg.1 |
TC17000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007604.hg.1 |
ENST00000322172.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007604.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007604.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007604.hg.1 |
TC17000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007610.hg.1 |
ENST00000322172.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007610.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007610.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007610.hg.1 |
TC17000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159963.hg.1 |
TC1700008144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159970.hg.1 |
TC1700008144.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008144.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
ENST00000376741.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
ENST00000613139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
NM_003110.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159968.hg.1 |
TC1700008144.hg.1 |
Clariom D TC |