Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
TC06004092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000274897.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000358276.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000370876.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000370877.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000370878.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000502396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000509997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000511744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
ENST00000514921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008042.hg.1 |
NM_138569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000274897.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000370876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000370877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000447836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000502396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000509997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000511744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000514433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000514921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_001281746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_001281747.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_138569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000274897.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000370876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000370877.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000447836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000502396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000509997.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000511744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000514433.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
ENST00000514921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_001281746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_001281747.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
NM_138569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198898.hg.1 |
TC0600014138.hg.1 |
Clariom D TC |