Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043379.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043379.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043379.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010637.hg.1 |
ENST00000561269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008279.hg.1 |
ENST00000513388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043376.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043376.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043376.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043376.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043380.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043380.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043380.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043374.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043374.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043374.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043374.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043377.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043377.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043377.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011417.hg.1 |
ENST00000550884.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043378.hg.1 |
TC02002684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043378.hg.1 |
ENST00000261015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043378.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000463824.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000464820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000467777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000475611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000477727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015463.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213633.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011417.hg.1 |
ENST00000550884.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010637.hg.1 |
ENST00000561269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213635.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213635.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213635.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213635.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213635.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000477723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
ENST00000478869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213631.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213636.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213636.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213636.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213636.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213636.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008279.hg.1 |
ENST00000513388.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213634.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213634.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213634.hg.1 |
ENST00000261015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213634.hg.1 |
NM_018256.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213634.hg.1 |
TC0200015463.hg.1 |
Clariom D TC |