Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011094.hg.1 |
ENST00000177742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011094.hg.1 |
ENST00000397375.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011094.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011094.hg.1 |
TC16000759.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011095.hg.1 |
ENST00000177742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011095.hg.1 |
ENST00000397375.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011095.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011095.hg.1 |
TC16000759.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009063.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000177742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000397375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000569585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
ENST00000613089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
NM_001300900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
NM_023936.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168654.hg.1 |
TC1600009063.hg.1 |
Clariom D TC |