Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011519152.2 | XP_011517454.1 | 12 - 13 | 1748 | 79 | |
XM_011519158.1 | XP_011517460.1 | 12 - 13 | 1417 | 79 | ||
GenBank mRNA | AF064105.2 | - | 12 - 13 | 1801 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons |