Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024139.hg.1 |
ENST00000221973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024139.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024139.hg.1 |
TC19001777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194715.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194715.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194715.hg.1 |
TC1900011252.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194715.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194715.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000221973.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
ENST00000596399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_001161748.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011252.hg.1 |
NM_030657.3 |
Refseq |