Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007248.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007248.hg.1 |
ENST00000379486.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007248.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007248.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007248.hg.1 |
TC16000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159891.hg.1 |
TC1600008126.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000379486.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000532362.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_001199317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008126.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
ENST00000268595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
ENST00000569316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
NM_144673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159889.hg.1 |
TC1600008126.hg.1 |
Clariom D TC |