Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007903.hg.1 |
ENST00000605170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019106.hg.1 |
ENST00000278200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019106.hg.1 |
ENST00000526776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019106.hg.1 |
ENST00000532287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019106.hg.1 |
NM_144981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019106.hg.1 |
TC11001525.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007903.hg.1 |
ENST00000605170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000278200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000526776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000527184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000529749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000530023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000531693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000532287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000532624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
NM_001304274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
NM_144981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000278200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000526776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000527184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000529749.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000530023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000531693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000532287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
ENST00000532624.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
NM_001304274.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
NM_144981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179202.hg.1 |
TC1100010451.hg.1 |
Clariom D TC |