Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010704.hg.1 |
TC06000916.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010704.hg.1 |
ENST00000319550.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010704.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000417846.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000419791.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009228.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
ENST00000319550.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
ENST00000460749.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
NM_152729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163334.hg.1 |
TC0600009228.hg.1 |
Clariom D TC |