Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004769.hg.1 |
ENST00000293599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004769.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004769.hg.1 |
TC12000384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153648.hg.1 |
TC1200007592.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200153646.hg.1 |
TC1200007592.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000293599.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
ENST00000553132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007592.hg.1 |
NM_001651.3 |
Refseq |