Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005198.hg.1 |
TC04000381.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005198.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005198.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153063.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153063.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153063.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153063.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153063.hg.1 |
TC0400007776.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
ENST00000317987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007776.hg.1 |
NM_152997.3 |
Refseq |