Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045518.hg.1 |
TC02002821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045518.hg.1 |
ENST00000233055.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045518.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000462702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000462702.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045517.hg.1 |
TC02002821.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045517.hg.1 |
ENST00000233055.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045517.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216716.hg.1 |
TC0200015870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000462702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000462702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000483061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015870.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000233055.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000429915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000491453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
ENST00000493001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
NM_020830.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200216717.hg.1 |
TC0200015870.hg.1 |
Clariom D TC |