Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014987.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014987.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014987.hg.1 |
TC10001194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014988.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014988.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014988.hg.1 |
TC10001194.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000435805.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000452075.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000435805.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000452075.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170739.hg.1 |
TC1000010408.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000435805.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000452075.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000435805.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000452075.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
ENST00000473592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
NR_026777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170747.hg.1 |
TC1000010408.hg.1 |
Clariom D TC |