Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013780.hg.1 |
TC02000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013780.hg.1 |
ENST00000288670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013780.hg.1 |
ENST00000421344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013780.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013780.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013781.hg.1 |
TC02000941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013781.hg.1 |
ENST00000288670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013781.hg.1 |
ENST00000421344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013781.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013781.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167669.hg.1 |
TC0200009680.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000492942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
ENST00000497192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009680.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
ENST00000288670.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
ENST00000475377.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
NM_052905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167668.hg.1 |
TC0200009680.hg.1 |
Clariom D TC |