Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1549L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012194.2 | NP_036326.2 | 15 - 16 | 4644 | 74 | |
XM_005252847.3 | XP_005252904.2 | 17 - 18 | 4738 | 74 | ||
XM_005252848.3 | XP_005252905.1 | 15 - 16 | 4659 | 74 | ||
XM_011519970.2 | XP_011518272.1 | 16 - 17 | 3304 | 74 | ||
XM_017017484.1 | XP_016872973.1 | 15 - 16 | 4662 | 74 | ||
XM_017017485.1 | XP_016872974.1 | 15 - 16 | 4662 | 74 | ||
XM_017017486.1 | XP_016872975.1 | 16 - 17 | 3301 | 74 | ||
GenBank mRNA | U10991.1 | - | 15 - 16 | 4051 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |