Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
ENST00000391762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
ENST00000391764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
ENST00000417328.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
ENST00000485876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
NM_004542.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011664.hg.1 |
TC19000871.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
TC1900011795.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900202544.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000303553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391763.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000391764.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000417903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000420296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000451517.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000471292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000484103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011795.hg.1 |
ENST00000485876.5 |
Ensembl |