Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
ENST00000402860.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
ENST00000405091.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22012658.hg.1 |
TC22000717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
TC2200008629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162239.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000381821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000402860.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000405091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
ENST00000442538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_001163857.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008629.hg.1 |
NM_178552.3 |
Refseq |